CAS RN 290315-45-6
IUPAC Name:
4-(2-{1-[N-(2,5-difluorophenyl)4-chlorobenzenesulfonamido]ethyl}-5-fluorophenyl)butanoic acid
Synonyms:
4-(2-{1-[N-(2,5-difluorophenyl)4-chlorobenzenesulfonamido]ethyl}-5-fluorophenyl)butanoic acid
Molecular Weight (g/mol):
511.94
Molecular Formula:
C24H21ClF3NO4S
InChi Key:
IZAOBRWCUGOKNH-UHFFFAOYNA-N
SMILES:
CC(N(C1=C(F)C=CC(F)=C1)S(=O)(=O)C1=CC=C(Cl)C=C1)C1=C(CCCC(O)=O)C=C(F)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ BMS 299897
CAS: 290315-45-6 Molecular Formula: C24H21ClF3NO4S