accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29342-05-0

IUPAC Name: 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
Synonyms: ciclopirox penlac loprox batrafen penlac nail lacquer stieprox ciclopiroxolamine loprox gel terit loprox shampoo
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: SCKYRAXSEDYPSA-UHFFFAOYSA-N
SMILES: CC1=CC(=O)N(O)C(=C1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results