accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNFFFSOOFFFFFF

CAS RN 293754-55-9

IUPAC Name: N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
Synonyms: N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
Molecular Weight (g/mol): 481.33
Molecular Formula: C17H12F9NO3S
InChi Key: SGIWFELWJPNFDH-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(C=C1)N(CC(F)(F)F)S(=O)(=O)C1=CC=CC=C1)(C(F)(F)F)C(F)(F)F
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)