CAS RN 2941-69-7
IUPAC Name:
5-methoxy-2-methyl-1,3-benzothiazole
Synonyms:
5-methoxy-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol):
179.24
Molecular Formula:
C9H9NOS
InChi Key:
SAQMNBWVOKYKPZ-UHFFFAOYSA-N
SMILES:
COC1=CC=C2SC(C)=NC2=C1
1
–
1
of
1
results
1
5-Methoxy-2-methylbenzothiazole 98.0+%, TCI America™
CAS: 2941-69-7 Molecular Formula: C9H9NOS Molecular Weight (g/mol): 179.24 MDL Number: MFCD00082664 InChI Key: SAQMNBWVOKYKPZ-UHFFFAOYSA-N PubChem CID: 76252 IUPAC Name: 5-methoxy-2-methyl-1,3-benzothiazole SMILES: COC1=CC=C2SC(C)=NC2=C1