accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 29734-68-7

IUPAC Name: (1S,16R)-16-methoxy-4-oxo-5-oxa-10-azatetracyclo[8.7.0.0¹,¹³.0²,⁷]heptadeca-2(7),14-dien-10-ium bromide
Synonyms: (1S,16R)-16-methoxy-4-oxo-5-oxa-10-azatetracyclo[8.7.0.0¹,¹³.0²,⁷]heptadeca-2(7),14-dien-10-ium bromide
Molecular Weight (g/mol): 356.26
Molecular Formula: C16H22BrNO3
InChi Key: ITZHVVLSHDXSIJ-WDXUOIDCSA-N
SMILES: [Br-].CO[C@@H]1C[C@@]23C(CC[NH+]2CCC2=C3CC(=O)OC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)