Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Dihydro-β-erythroidine hydrobromide
GREENER_CHOICESDP

Catalog No. 234950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-495-0 50 mg
23-491-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-495-0 Supplier Tocris Bioscience™ Supplier No. 2349/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α4β2, muscle type and Torpedo nAChR antagonist

Dihydro-β-erythroidine hydrobromide is a member of the Erythrina alkaloids, it is a competitive nicotinic acetylcholine receptor antagonist with moderate selectivity for the neuronal α4 receptor subunit (IC50 values are 0.19 and 0.37 μM for α4β4 and α4β2 receptors, respectively). Antagonizes behavioral effects of nicotine in vivo. Dihydro-β-erythroidine blocks excitation of striatal GABAergic neurons, which completely suppress polysynaptic inhibition between striatal cholinergic interneurons. Dihydro-β-erythroidine hydrobromide has antidepressive-like effects in mice (forced swim and mouse suspension). Orally bioavailable

Chemical Identifiers

CAS 29734-68-7
Molecular Formula C16H22BrNO3

Specifications

Quantity 50 mg
Formula Weight 356.26
Purity >98%
Molecular Weight (g/mol) 356.26
Chemical Name or Material Dihydro-β-erythroidine hydrobromide
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.