accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 300-38-9

IUPAC Name: (2S)-2-amino-3-(3,5-dibromo-4-hydroxyphenyl)propanoic acid
Synonyms: 3,5 dibromotyrosine dibromotyrosine 二溴酪氨酸 bromotiren DiBrY dibromotirina C9H9Br2NO3 biotiren
Molecular Weight (g/mol): 338.98
Molecular Formula: C9H9Br2NO3
InChi Key: COESHZUDRKCEPA-ZETCQYMHSA-N
SMILES: N[C@@H](CC1=CC(Br)=C(O)C(Br)=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results