accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ClOHNNS

CAS RN 300851-67-6

IUPAC Name: 2-(4-chlorophenyl)-3-methyl-N-(1,3-thiazol-2-yl)butanamide
Synonyms: 2-(4-chlorophenyl)-3-methyl-N-(1,3-thiazol-2-yl)butanamide
Molecular Weight (g/mol): 294.8
Molecular Formula: C14H15ClN2OS
InChi Key: AZYDQCGCBQYFSE-UHFFFAOYNA-N
SMILES: CC(C)C(C(=O)NC1=NC=CS1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)