CAS RN 301326-22-7
IUPAC Name:
1-methyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}-1H-pyrazole-5-carboxamide
Synonyms:
2-methyl-N-{2-methyl-4-[2-(2-methylphenyl)diazen-1-yl]phenyl}pyrazole-3-carboxamide
Molecular Weight (g/mol):
333.4
Molecular Formula:
C19H19N5O
InChi Key:
LKTNEXPODAWWFM-UHFFFAOYSA-N
SMILES:
CN1N=CC=C1C(=O)NC1=C(C)C=C(C=C1)N=NC1=CC=CC=C1C
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ AhR Antagonist
The AhR Antagonist, also referenced under CAS 301326-22-7, controls the biological activity of AhR.
2
R&D Systems™ CH 223191
CAS: 301326-22-7 Molecular Formula: C19H19N5O