Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CH 223191
SDP

Catalog No. 385850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-585-0 50 mg
38-581-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-585-0 Supplier R&D Systems™ Supplier No. 3858/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent aryl hydrocarbon receptor (AhR) antagonist

Potent aryl hydrocarbon receptor (AhR) antagonist (IC50 = 30 nM). Exhibits no AhR agonist-like activity (at concentrations up to 100 μM). Inhibits 2,3,7,8-Tetrachlorodibenzo-p-dioxin (TCDD)-induced AhR-dependent transcription in vitro and reduces TCDD-induced toxicity in vivo. Attenuates Th17 differentiation of naive CD4 T cells and promotes expansion of hematopoietic stem cells in vitro.

Chemical Identifiers

CAS 301326-22-7
Molecular Formula C19H19N5O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 333.39
Purity >98%
Molecular Weight (g/mol) 333.395
Chemical Name or Material CH 223191
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.