accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHNOH2NN

CAS RN 301836-43-1

IUPAC Name: 4-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl]benzamide
Synonyms: 4-[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-(pyridin-2-yl)-1H-imidazol-2-yl]benzamide
Molecular Weight (g/mol): 398.42
Molecular Formula: C23H18N4O3
InChi Key: DPDZHVCKYBCJHW-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=N1)C1=CC2=C(OCCO2)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)