accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3OOCH3CH3

CAS RN 3039-71-2

IUPAC Name: hydrogen 8-[2-(diethylamino)ethoxy]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one chloride
Synonyms: hydrogen 8-[2-(diethylamino)ethoxy]-9a,11a-dimethyl-2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9bH,10H,11H-cyclopenta[a]phenanthren-1-one chloride
Molecular Weight (g/mol): 424.07
Molecular Formula: C25H42ClNO2
InChi Key: ICPRVJHNLABCSA-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCN(CC)CCOC1CCC2=CCC3C4CCC(=O)C4(C)CCC3C2(C)C1
Product Type 
  • (1)
  • (2)
Form 
  • (1)
Product Line 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results