Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U 18666A, R&D Systems™
SDP

Catalog No. 163810 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-381-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-381-0 Supplier R&D Systems Supplier No. 1638/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of Hedgehog (Hh) signaling. Also inhibits cholesterol synthesis

Inhibitor of cholesterol synthesis and cellular transport, and weak inhibitor of hedgehog (Hh) signaling. Reduces serum sterol levels in rats in vivo.

Chemical Identifiers

CAS 3039-71-2
Molecular Formula C25H42ClNO2
Molecular Weight (g/mol) 424.07
MDL Number MFCD00210908
InChI Key ICPRVJHNLABCSA-UHFFFAOYNA-N
Synonym unii-v0dpr5j8zj, v0dpr5j8zj, 3beta-2-diethylaminoethoxy androst-5-en-17-one, hcl, 3beta-2-diethylaminoethoxy androst-5-en-17-one hydrochloride, u 18666a, powder, dsstox_cid_26904, dsstox_rid_82003, dsstox_gsid_46904, androst-5-en-17-one, hydrochloride, androst-5-en-17-one, hydrochloride, 3.beta.
PubChem CID 9954082
IUPAC Name hydrogen 8-[2-(diethylamino)ethoxy]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one chloride
SMILES [H+].[Cl-].CCN(CC)CCOC1CCC2=CCC3C4CCC(=O)C4(C)CCC3C2(C)C1

Specifications

Quantity 10 mg
Formula Weight 424.07
Percent Purity >98%
Product Line Tocris
Chemical Name or Material U 18666A
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.