CAS RN 304448-55-3
IUPAC Name:
3-hydroxy-N'-[(3-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]naphthalene-2-carbohydrazide
Synonyms:
3-hydroxy-N'-[(3-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]naphthalene-2-carbohydrazide
Molecular Weight (g/mol):
322.32
Molecular Formula:
C18H14N2O4
InChi Key:
NRQJUIHYFNUXFT-UHFFFAOYSA-N
SMILES:
OC1=CC(=CNNC(=O)C2=CC3=CC=CC=C3C=C2O)C=CC1=O
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Dynamin Inhibitor I, Dynasore
The Dynamin Inhibitor I, Dynasore, also referenced under CAS 304448-55-3, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.
2
R&D Systems™ Dynasore
CAS: 304448-55-3 Molecular Formula: C18H14N2O4