Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Dynasore
GREENER_CHOICESDP

Catalog No. 289750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-975-0 50 mg
28-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-975-0 Supplier R&D Systems™ Supplier No. 2897/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Non-competitive dynamin inhibitor

Dynasore is a non-competitive reversible inhibitor of dynamin 1, dynamin 2 and mitochondrial dynamin (Drp1) GTPase activity. Dynasore blocks endocytic pathways dependent on dynamin and inhibits cell spreading and migration of BSC1 cells. Dynasore attenuates motor symptoms in an animal model of Parkinson's disease and in a cellular model of Alzheimer's disease it reduces tau protein aggregation and internalization.

Chemical Identifiers

CAS 304448-55-3
Molecular Formula C18H14N2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 322.32
Purity >99%
Molecular Weight (g/mol) 322.32
Chemical Name or Material Dynamin Inhibitors
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.