accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOBrI

CAS RN 304679-75-2

IUPAC Name: 2-[2-(4-bromophenyl)-2-oxoethyl]-1-methylpyridin-1-ium iodide
Synonyms: 2-[2-(4-bromophenyl)-2-oxoethyl]-1-methylpyridin-1-ium iodide
Molecular Weight (g/mol): 418.07
Molecular Formula: C14H13BrINO
InChi Key: LERUHBBUGAOYOD-UHFFFAOYSA-M
SMILES: [I-].C[N+]1=CC=CC=C1CC(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)