accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3NNN(E/Z)

CAS RN 304909-07-7

IUPAC Name: N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methanimine
Synonyms: N-(4-benzylpiperazin-1-yl)-1-(3,5-dimethyl-1-phenylpyrazol-4-yl)methanimine
Molecular Weight (g/mol): 373.5
Molecular Formula: C23H27N5
InChi Key: FOORCIAZMIWALX-UHFFFAOYSA-N
SMILES: CC1=NN(C(C)=C1C=NN1CCN(CC2=CC=CC=C2)CC1)C1=CC=CC=C1
Product Type 
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results