accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHHONSOOONHOOCH3

CAS RN 307002-73-9

IUPAC Name: N-[2-(4-{benzyl[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl}phenoxy)ethyl]-1-benzofuran-2-carboxamide
Synonyms: N-[2-(4-{benzyl[2-(hydroxycarbamoyl)-4,6-dimethylphenyl]sulfamoyl}phenoxy)ethyl]-1-benzofuran-2-carboxamide
Molecular Weight (g/mol): 613.69
Molecular Formula: C33H31N3O7S
InChi Key: FARMEEAGJWMFSZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N(CC2=CC=CC=C2)S(=O)(=O)C2=CC=C(OCCNC(=O)C3=CC4=CC=CC=C4O3)C=C2)C(=C1)C(=O)NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)