accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 307510-92-5

OHOSSNOFFF(Z)

CAS RN 307510-92-5

IUPAC Name: 4-{[(5Z)-4-oxo-2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-5-ylidene]methyl}benzoic acid
Synonyms: 4-{[(5Z)-4-oxo-2-sulfanylidene-3-[3-(trifluoromethyl)phenyl]-1,3-thiazolidin-5-ylidene]methyl}benzoic acid
Molecular Weight (g/mol): 409.4
Molecular Formula: C18H10F3NO3S2
InChi Key: JIMHYXZZCWVCMI-ZSOIEALJSA-N
SMILES: OC(=O)C1=CC=C(\C=C2/SC(=S)N(C2=O)C2=CC=CC(=C2)C(F)(F)F)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results