accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 307538-42-7

IUPAC Name: 6-bromo-N-(prop-2-en-1-yl)quinazolin-4-amine
Synonyms: 6-bromo-N-(prop-2-en-1-yl)quinazolin-4-amine
Molecular Weight (g/mol): 264.13
Molecular Formula: C11H10BrN3
InChi Key: BCPOLXUSCUFDGE-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NCC=C)N=CN=C2C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results