accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4H3CNSSOOONNSNH3CSOOO(Z)(Z)

CAS RN 30931-67-0

IUPAC Name: diammonium (2Z)-3-ethyl-2-{2-[(2Z)-3-ethyl-6-sulfonato-2,3-dihydro-1,3-benzothiazol-2-ylidene]hydrazin-1-ylidene}-2,3-dihydro-1,3-benzothiazole-6-sulfonate
Synonyms: ABTS
Molecular Weight (g/mol): 548.67
Molecular Formula: C18H24N6O6S4
InChi Key: OHDRQQURAXLVGJ-AXMZSLBLSA-N
SMILES: [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O
Quantity 
  • (1)
  • (5)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)