Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ABTS™ Chromophore, Diammonium Salt, Calbiochem™,

Catalog No. 1944301GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
19-443-01GM 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-443-01GM Supplier MilliporeSigma™ Supplier No. 1944301GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Substrate useful for ELISA that acts as a hydrogen donor in peroxidase reactions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
Molecular Weight (g/mol) 548.67
MDL Number MFCD00010404,MFCD00010404
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
PubChem CID 91884754
IUPAC Name diammonium (2Z)-3-ethyl-2-{2-[(2Z)-3-ethyl-6-sulfonato-2,3-dihydro-1,3-benzothiazol-2-ylidene]hydrazin-1-ylidene}-2,3-dihydro-1,3-benzothiazole-6-sulfonate
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O

Specifications

Color Green
Packaging Glass Bottle
Quantity 1 g
Percent Purity ≥98%
Grade Not Available
Physical Form Crystalline Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.