accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOClHNOHNNNN

CAS RN 312637-48-2

IUPAC Name: N-[3-(4-chlorophenyl)-1-{4-cyclohexyl-4-[(1H-1,2,4-triazol-1-yl)methyl]piperidin-1-yl}-1-oxopropan-2-yl]-1,2,3,4-tetrahydroisoquinoline-3-carboxamide
Synonyms: THIQ
Molecular Weight (g/mol): 589.18
Molecular Formula: C33H41ClN6O2
InChi Key: HLCHESOMJVGDSJ-UHFFFAOYNA-N
SMILES: ClC1=CC=C(CC(NC(=O)C2CC3=CC=CC=C3CN2)C(=O)N2CCC(CN3C=NC=N3)(CC2)C2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results