Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ THIQ
GREENER_CHOICESDP

Catalog No. 30321 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-321 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 30-321 Supplier R&D Systems™ Supplier No. 3032/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective MC4 receptor agonist

Potent and selective melanocortin 4 (MC4) receptor agonist (IC50 values are 1.2, 761 and 2067 nM for human MC4, MC3 and MC1 receptors respectively). Enhances intracavernosal pressure and stimulates erectile activity in rats ex copula following systemic administration.

Specifications

CAS 312637-48-2
Quantity 1 mg
Product Line Tocris
Product Type THIQ
Purity >95%
Molecular Weight (g/mol) 589.181
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.