CAS RN 312756-74-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ RAD51-Stimulatory Compound-1, RS-1
A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis.
2
Tocris Bioscience™ RS 1
High affinity RAD51 activator; also CRISPR enhancer