accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNSOONHNS

CAS RN 313254-51-2

IUPAC Name: 2,2-diphenyl-N-{4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}acetamide
Synonyms: 2,2-diphenyl-N-{4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}acetamide
Molecular Weight (g/mol): 449.54
Molecular Formula: C23H19N3O3S2
InChi Key: URSQNPPONHUJDL-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1)C(C1=CC=CC=C1)C1=CC=CC=C1
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)