Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ICA 121431
GREENER_CHOICESDP

Catalog No. 506650R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
506650R 50 mg
506610 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 506650R Supplier R&D Systems™ Supplier No. 5066/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective NaV1.3 and NaV1.1 channel blocker

Potent and selective inhibitor of human NaV1.3 and NaV1.1 channels (IC50 values are 13 and 23 nM respectively). Exhibits up to 1,000 fold selectivity against other TTX-sensitive or resistant sodium channels (IC50 values are >10 μM for human NaV1.5 and NaV1.8 channels). Interacts with an inhibitory interaction site distinct from those bound by TTX and local anesthetic-like modulators.

Chemical Identifiers

CAS 313254-51-2
Molecular Formula C23H19N3O3S2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 449.55
Physical Form Solid
Color White
Purity >98%
Molecular Weight (g/mol) 449.543
Chemical Name or Material ICA 121431
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.