accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOONHNCl

CAS RN 313516-66-4

IUPAC Name: 2-chloro-5-nitro-N-(pyridin-4-yl)benzamide
Synonyms: 2-chloro-5-nitro-N-(pyridin-4-yl)benzamide
Molecular Weight (g/mol): 277.66
Molecular Formula: C12H8ClN3O3
InChi Key: FRPJSHKMZHWJBE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(C(=O)NC2=CC=NC=C2)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results