Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ T 0070907
GREENER_CHOICESDP

Catalog No. 23015 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-015 5 mg
23-012-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-015 Supplier Tocris Bioscience™ Supplier No. 2301/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent and selective covalent PPARγ antagonist

T 0070907 is a potent and selective irreversible PPARγ antagonist (IC50 = 1 nM). Displays > 800-fold selectivity for PPARγ over PPARα and PPARδ. Blocks transcriptional activity of PPARγ in vitro and inhibits Rosiglitazone -induced adipogenesis.

Specifications

Chemical Name or Material 2-Chloro-5-nitro-N-4-pyridinylbenzamide
CAS 313516-66-4
Assay Percent Range >99%
Quantity 5 mg
Molecular Formula C12H8N3O3Cl
Formula Weight Observed MW: 277.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.