Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Evans Blue tetrasodium salt
GREENER_CHOICESDP

Catalog No. 08451G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-451-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-451-G Supplier R&D Systems™ Supplier No. 0845/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dye for assessing cell viability and blood brain barrier permeability; also EAAT inhibitor and iGluR antagonist

Potent inhibitor of L-glutamate uptake into synaptic vesicles. Also inhibits AMPA and kainate receptor-mediated currents (IC50 values are 220 and 150 nM respectively). P2X-selective purinoceptor antagonist. Also a non-permeating dye, stains damaged plasma membrane and enters in the cytoplasm and nucleus, used for checking cell viability.

Chemical Identifiers

CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 960.82
Purity >95%
Molecular Weight (g/mol) 960.79
Chemical Name or Material Evans Blue tetrasodium salt
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.