CAS RN 314-19-2
IUPAC Name:
hydrogen (9R)-10-methyl-10-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,13,15-hexaene-3,4-diol chloride
Synonyms:
apomorphine hydrochloride
Molecular Weight (g/mol):
303.79
Molecular Formula:
C17H18ClNO2
InChi Key:
SKYZYDSNJIOXRL-BTQNPOSSSA-N
SMILES:
[H+].[Cl-].CN1CCC2=CC=CC3=C2[C@H]1CC1=CC=C(O)C(O)=C31
1
–
1
of
1
results
1
R&D Systems™ (R)-(-)-Apomorphine hydrochloride
CAS: 314-19-2 Molecular Formula: C17H18ClNO2