CAS RN 314045-39-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Glutaminase Inhibitor II, BPTES
A cell-permeable, potent, selective, reversible, and uncompetitive allosteric inhibitor of kidney-type glutaminase (GLS1). Does not affect the activity of liver glutaminase.
2
Tocris Bioscience™ BPTES
Glutaminase Inhibitors