accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 314054-36-9

IUPAC Name: N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-2-oxopiperidine-3-carboxamide
Synonyms: HIOC
Molecular Weight (g/mol): 301.35
Molecular Formula: C16H19N3O3
InChi Key: ZIMKJLALTRLXJO-UHFFFAOYNA-N
SMILES: OC1=CC=C2NC=C(CCNC(=O)C3CCCNC3=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results