Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HIOC, >98%
GREENER_CHOICESDP

Catalog No. 596150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-615-0 50 mg
59-611-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-615-0 Supplier Tocris Bioscience™ Supplier No. 5961/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective TrkB receptor agonist; neuroprotectant

Selective TrkB receptor agonist. Protects neurons from excitotoxicity. Protects retinas from light-induced retinal degeneration (LIRD) in vivo. Blood brain barrier and blood-retinal barrier penetrant.

Chemical Identifiers

CAS 314054-36-9
Molecular Formula C16H19N3O3

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 301.34
Purity >98%
Molecular Weight (g/mol) 301.346
Chemical Name or Material N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]-2-oxo-3-piperidinecarboxamide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.