accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31491-21-1

H3CSiH3CCH3OOSiCH3H3CCH3

CAS RN 31491-21-1

IUPAC Name: 2,2,6,6-tetramethyl-4-(phenylmethylidene)-3,5-dioxa-2,6-disilaheptane
Synonyms: 2,2,6,6-tetramethyl-4-(phenylmethylidene)-3,5-dioxa-2,6-disilaheptane
Molecular Weight (g/mol): 280.51
Molecular Formula: C14H24O2Si2
InChi Key: HMHSNGLUZWVXQY-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(O[Si](C)(C)C)=CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results