CAS RN 315694-89-4
IUPAC Name:
2-(2-phenylethenyl)-4-[(pyridin-3-yl)methylidene]-4,5-dihydro-1,3-oxazol-5-one
Synonyms:
2-(2-phenylethenyl)-4-(pyridin-3-ylmethylidene)-1,3-oxazol-5-one
Molecular Weight (g/mol):
276.3
Molecular Formula:
C17H12N2O2
InChi Key:
GFGMISOSPOPSHN-UHFFFAOYSA-N
SMILES:
O=C1OC(C=CC2=CC=CC=C2)=NC1=CC1=CN=CC=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ DAPK Inhibitor
The DAPK Inhibitor, also referenced under CAS 315694-89-4, controls the biological activity of DAPK.
2
R&D Systems™ TC-DAPK 6
CAS: 315694-89-4 Molecular Formula: C17H12N2O2