accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 31576-33-7

H3CCH3OOCH3

CAS RN 31576-33-7

IUPAC Name: 4-(2-methylpropoxy)butan-2-one
Synonyms: 4-(2-methylpropoxy)butan-2-one
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: VDVQAHFUWSAGRT-UHFFFAOYSA-N
SMILES: CC(C)COCCC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results