accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2CCH3H3COHOH

CAS RN 317321-41-8

IUPAC Name: 5-methyl-4-[3-methyl-6-(prop-1-en-2-yl)cyclohex-2-en-1-yl]benzene-1,3-diol
Synonyms: 5-methyl-4-[3-methyl-6-(prop-1-en-2-yl)cyclohex-2-en-1-yl]benzene-1,3-diol
Molecular Weight (g/mol): 258.36
Molecular Formula: C17H22O2
InChi Key: KDZOUSULXZNDJH-UHFFFAOYNA-N
SMILES: CC(=C)C1CCC(C)=CC1C1=C(O)C=C(O)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results