Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ O-1602
GREENER_CHOICESDP

Catalog No. 279710 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-971-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-971-0 Supplier R&D Systems™ Supplier No. 2797/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent GPR55 agonist

Analog of cannabidiol that is a potent agonist at the GPR55 cannabinoid receptor (EC50 values are 13, > 30000 and > 30000 nM for GPR55, CB1 and CB2 receptors respectively). Induces activation of RhoA, cdc42 and rac1. Increases proliferation of neural stem cells NSCs in vitro and in vivo. Also promotes neurogenesis in vivo.

Chemical Identifiers

CAS 317321-41-8
Molecular Formula C17H22O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 258.36
Purity >98%
Molecular Weight (g/mol) 258.361
Chemical Name or Material O-1602
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.