accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOHOOH3C

CAS RN 3187-19-7

IUPAC Name: 2-hydroxy-4,6-dimethoxybenzoic acid
Synonyms: 2-hydroxy-4,6-dimethoxybenzoic acid
Molecular Weight (g/mol): 198.17
Molecular Formula: C9H10O5
InChi Key: LEIJLTBRQHFMDL-UHFFFAOYSA-N
SMILES: COC1=CC(O)=C(C(O)=O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results