CAS RN 319460-85-0
IUPAC Name:
N-methyl-2-({3-[(1E)-2-(pyridin-2-yl)ethenyl]-1H-indazol-6-yl}sulfanyl)benzamide
Synonyms:
axitinib
Molecular Weight (g/mol):
386.47
Molecular Formula:
C22H18N4OS
InChi Key:
RITAVMQDGBJQJZ-FMIVXFBMSA-N
SMILES:
CNC(=O)C1=CC=CC=C1SC1=CC=C2C(NN=C2\C=C\C2=CC=CC=N2)=C1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Axitinib
Thermo Scientific Chemicals Axitinib
CAS: 319460-85-0 Molecular Formula: C22H18N4OS Molecular Weight (g/mol): 386.47 InChI Key: RITAVMQDGBJQJZ-FMIVXFBMSA-N IUPAC Name: N-methyl-2-({3-[(1E)-2-(pyridin-2-yl)ethenyl]-1H-indazol-6-yl}sulfanyl)benzamide SMILES: CNC(=O)C1=CC=CC=C1SC1=CC=C2C(NN=C2\C=C\C2=CC=CC=N2)=C1
2
Tocris Bioscience™ Axitinib
CAS: 319460-85-0 Molecular Formula: C22H18N4OS