Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Axitinib
GREENER_CHOICESDP

Catalog No. 435050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-505-0 50 mg
43-501-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-505-0 Supplier R&D Systems™ Supplier No. 4350/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent VEGFR-1, -2 and -3 inhibitor

Axitinib is a potent inhibitor of VEGFR-2, -3, and -1 (IC50 values are 0.2, 0.1-0.3, and 1.2 nM respectively). Axitinib exhibits minimal activity against a panel of ∼100 protein kinases. Axitinib inhibits angiogenesis and vascular permeability. Also a high affinity BCR-ABL1 (T315I) inhibitor (Ki = 149 pM for autophosphorylated ABL1 (T315I)). Axitinib blocks proliferation of Ba/F3 cells expressing BCR-ABL1 (T315I). Axitinib (10 mg/kg) decreases T-cell exhaustion and tumor and T-cell hypoxia and improves survival in mice in vivo. Orally available.

Chemical Identifiers

CAS 319460-85-0
Molecular Formula C22H18N4OS

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 386.47
Purity >99%
Molecular Weight (g/mol) 386.473
Chemical Name or Material Axitinib
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.