accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32385-58-3

IUPAC Name: {[2-(1H-imidazol-5-yl)ethyl]sulfanyl}methanimidamide dihydrobromide
Synonyms: imetit dihydrobromide
Molecular Weight (g/mol): 332.06
Molecular Formula: C6H12Br2N4S
InChi Key: DOBOYMKCRRLTRF-UHFFFAOYSA-N
SMILES: Br.Br.NC(=N)SCCC1=CN=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results