Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Imetit dihydrobromide
GREENER_CHOICESDP

Catalog No. 072950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-295-0 50 mg
07-291-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-295-0 Supplier R&D Systems™ Supplier No. 0729/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Standard H3 and H4 agonist (H3 > H4)

An extremely potent, high affinity agonist at H3 and H4 receptors (Ki values are 0.3 and 2.7 nM respectively). Induces shape change in eosinophils with an EC50 of 25 nM. Centrally active following systemic administration.

Chemical Identifiers

CAS 32385-58-3
Molecular Formula C6H12Br2N4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 332.06
Purity >98%
Molecular Weight (g/mol) 332.06
Chemical Name or Material Imetit dihydrobromide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.