CAS RN 325465-93-8
IUPAC Name:
sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Synonyms:
sodium 3-(hydrogen phosphonatooxy)-2-hydroxypropyl octadec-9-enoate
Molecular Weight (g/mol):
458.51
Molecular Formula:
C21H40NaO7P
InChi Key:
XGRLSUFHELJJAB-UHFFFAOYNA-M
SMILES:
[Na+].CCCCCCCCC=CCCCCCCCC(=O)OCC(O)COP(O)([O-])=O
1
–
1
of
1
results
1
R&D Systems™ 1-Oleoyl lysophosphatidic acid sodium salt
CAS: 325465-93-8 Molecular Formula: C21H40NaO7P