accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH2OHOOHO(Z)

CAS RN 32795-47-4

IUPAC Name: (2Z)-but-2-enedioic acid; 2-methyl-4-phenyl-1,2,3,4-tetrahydroisoquinolin-8-amine
Synonyms: maleic acid; nomifensine
Molecular Weight (g/mol): 354.41
Molecular Formula: C20H22N2O4
InChi Key: GEOCVSMCLVIOEV-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CN1CC(C2=CC=CC=C2)C2=CC=CC(N)=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)