accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOHO

CAS RN 328-50-7

IUPAC Name: 2-oxopentanedioic acid
Synonyms: oxoglutarate 2-oxyglutaric acid oxoglutaric acid ion(2-) 2-oxopentanedioic acid calcium ketoglutarate bis(L-arginin)-2-oxoglutarat α ketoglutarate (substance) alphaketoglutaric acid ketoglutaric acid
Molecular Weight (g/mol): 146.1
Molecular Formula: C5H6O5
InChi Key: KPGXRSRHYNQIFN-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)C(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (7)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)