CAS RN 328541-79-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ CFTR Inhibitor II, GlyH-101
CFTR Inhibitor II, GlyH-101, CAS 328541-79-3, is a cell-permeable potent, selective and reversible open-channel blocker of CFTR (Ki = 4.3 μM in CFTR-expressing FRT cells).
2
Tocris Bioscience™ GlyH 101
Reversible, voltage-dependent CFTR chloride channel blocker