CAS RN 330161-87-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ SU6656
A potent, cell-permeable, reversible, and ATP-competitive Src family kinase inhibitor.
2
Tocris Bioscience™ SU 6656
Potent ATP-competitive Src family kinase inhibitor (IC50 values are 0.02, 0.13, 0.17, 0.28 and 6.88 nM for Yes, Lyn, Fyn, Src and Lck, respectively)