accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHHNOBrSNHN(E/Z)

CAS RN 330676-02-3

IUPAC Name: 2-(1H-1,3-benzodiazol-2-ylsulfanyl)-N'-[(3-bromo-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]propanehydrazide
Synonyms: 2-(1H-1,3-benzodiazol-2-ylsulfanyl)-N'-[(3-bromo-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]propanehydrazide
Molecular Weight (g/mol): 419.3
Molecular Formula: C17H15BrN4O2S
InChi Key: GEHVZUUHMWNMAY-UHFFFAOYNA-N
SMILES: CC(SC1=NC2=CC=CC=C2N1)C(=O)NNC=C1C=C(Br)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)